4-Isoxazolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 4-Isoxazoleboronic acid pinacol ester

PN-8875-1G PN-8875-250M
COBBPN-8875-1GEA 324 EUR
COBBPN-8875-1G COBBPN-8875-250M
4-Isoxazolboronsäurepinakolester
4-Isoxazolboronsäurepinakolester
Formel: C₉H₁₄BNO₃
Molecular Weight: 195,02 g/mol
MDL: MFCD06657891
CAS-Nummer: 928664-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR