4-Bromisoxazol, Sigma-Aldrich®

Lieferant: Merck

724637-5G 724637-1G
SIAL724637-5GEA 74.8 EUR
SIAL724637-5G SIAL724637-1G
4-Bromisoxazol, Sigma-Aldrich®
4-Bromoisoxazole
Formel: C₃H₂BrNO
Molecular Weight: 147,9600 g/mol
MDL: MFCD02179569
CAS-Nummer: 97925-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR