Methylisoxazol-5-carboxylat ≥97%

Lieferant: Thermo Fisher Scientific

B25169.03 B25169.06
B25169.03EA 60.8 EUR
B25169.03 B25169.06
Methylisoxazol-5-carboxylat ≥97%
Methylisoxazol-5-carboxylat

Purity: 97% CAS No.: 15055-81-9 Molecular Formula: C5H5NO3 Formula Weight: 127.10 Melting Point: 48-50° MDL No.: MFCD01570794

Formel: C₅H₅NO₃
Molecular Weight: 127.10 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01570794
CAS-Nummer: 15055-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR