5-Amino-3-methylisoxazol-4-carbonitril

Lieferant: BIONET RESEARCH

3P-382S.500 3P-382S.1 3P-382S.5 3P-382S.10
BIOT3P-382S.500EA 0 EUR
BIOT3P-382S.500 BIOT3P-382S.1 BIOT3P-382S.5 BIOT3P-382S.10
5-Amino-3-methylisoxazol-4-carbonitril
5-Amino-3-methylisoxazol-4-carbonitril
Formel: C₅H₅N₃O
MDL: MFCD00027378
CAS-Nummer: 35261-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR