5-Amino-3-methylisoxazol-4-carbonitril

Lieferant: COMBI-BLOCKS

HI-1265-5G HI-1265-1G
COBBHI-1265-5GEA 530 EUR
COBBHI-1265-5G COBBHI-1265-1G
5-Amino-3-methylisoxazol-4-carbonitril
5-Amino-3-methylisoxazol-4-carbonitril
Formel: C₅H₅N₃O
MDL: MFCD00027378
CAS-Nummer: 35261-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR