Ethyl-5-methylisoxazol-3-carboxylat

Lieferant: Thermo Scientific

Synonyme: Ethyl 5-methylisoxazole-3-carboxylate

SPB01195.1 SPB01195.10
MAYBSPB01195.1EA 0 EUR
MAYBSPB01195.1 MAYBSPB01195.10
Ethyl-5-methylisoxazol-3-carboxylat
Ethyl-5-methylisoxazol-3-carboxylat
Formel: C₇H₉NO₃
Molecular Weight: 155,15 g/mol
MDL: MFCD00085080
CAS-Nummer: 3209-72-1
EINECS: 221-720-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR