Ethyl-5-methylisoxazol-3-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 5-methylisoxazole-3-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

681024-10G 681024-1G
SIAL681024-10GEA 268 EUR
SIAL681024-10G SIAL681024-1G
Ethyl-5-methylisoxazol-3-carboxylat, Sigma-Aldrich®
Ethyl-5-methylisoxazol-3-carboxylat
Formel: C₇H₉NO₃
Molecular Weight: 155,15 g/mol
MDL: MFCD00085080
CAS-Nummer: 3209-72-1
EINECS: 221-720-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR