Methyl-5-methylisoxazol-3-carboxylat ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl 5-methylisoxazole-3-carboxylate

A19814.22 A19814.14
A19814.22EA 309 EUR
A19814.22 A19814.14
Methyl-5-methylisoxazol-3-carboxylat ≥97%
Methyl-5-methylisoxazol-3-carboxylat

Purity: 97% CAS No.: 19788-35-3 Molecular Formula: C6H7NO3 Formula Weight: 141.13 Melting Point: 91-92° Boiling Point: 125°/11mm MDL No.: MFCD00015895

Formel: C₆H₇NO₃
Molecular Weight: 141.13 g/mol
Siedepunkt: 125 °C (11 mmHg)
Schmelzpunkt: 91…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015895
CAS-Nummer: 19788-35-3
EINECS: 243-311-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR