Ethyl-5-methylisoxazol-4-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 5-methylisoxazole-4-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

681016-25G 681016-5G
SIAL681016-25GEA 219 EUR
SIAL681016-25G SIAL681016-5G
Ethyl-5-methylisoxazol-4-carboxylat, Sigma-Aldrich®
Ethyl-5-methylisoxazol-4-carboxylat
Formel: C₇H₉NO₃
Molecular Weight: 155.15 g/mol
MDL: MFCD01631119
CAS-Nummer: 51135-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR