3,5-Dimethyl-4-isoxazolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
PN-3939-5G
PN-3939-1G
COBBPN-3939-5GEA
50.9
EUR
COBBPN-3939-5G
COBBPN-3939-1G
3,5-Dimethyl-4-isoxazolboronsäurepinakolester
3,5-Dimethyl-4-isoxazolboronsäurepinakolester
Formel:
C₁₁H₁₈BNO₃ Molecular Weight: 223.08 g/mol |
MDL:
MFCD05863910 CAS-Nummer: 832114-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...