3,5-Dimethyl-4-isoxazolboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
M-Clarity™ Qualitätsstufe = MQ100
643882-1G
643882-5G
SIAL643882-1GEA
26
EUR
SIAL643882-1G
SIAL643882-5G
3,5-Dimethyl-4-isoxazolboronsäurepinakolester, Sigma-Aldrich®
3,5-Dimethyl-4-isoxazolboronsäurepinakolester
Formel:
C₁₁H₁₈BNO₃ Molecular Weight: 223,08 g/mol |
MDL:
MFCD05863910 CAS-Nummer: 832114-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...