3-(Chlormethyl)-5-methylisoxazol

Lieferant: FLUOROCHEM

CFF272.1 CFF272.2.5 017319-1G 017319-2.5G
CHMPCFF272.1EA 348 EUR
CHMPCFF272.1 CHMPCFF272.2.5 FLUO017319-1G FLUO017319-2.5G
3-(Chlormethyl)-5-methylisoxazol
3-(Chloromethyl)-5-methylisoxazole
Formel: C₅H₆ClNO
Molecular Weight: 131.560 g/mol
MDL: MFCD00221166
CAS-Nummer: 35166-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR