3-(Hydroxymethyl)-5-methylisoxazol

Lieferant: FLUOROCHEM

Synonyme: 3-Hydroxymethyl-5-methylisoxazole, 5-Methyl-3-isoxazolemethanol

CFF465.2.5 CFF465.5 CFF465.10 017477-2.5G 017477-5G 017477-10G 017477-1G
CHMPCFF465.2.5EA 481 EUR
CHMPCFF465.2.5 CHMPCFF465.5 CHMPCFF465.10 FLUO017477-2.5G FLUO017477-5G FLUO017477-10G FLUO017477-1G
3-(Hydroxymethyl)-5-methylisoxazol
3-(Hydroxymethyl)-5-methylisoxazole
Formel: C₅H₇NO₂
Molecular Weight: 113,11 g/mol
MDL: MFCD00085129
CAS-Nummer: 35166-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR