4-Brom-2-(5-isoxazolyl)phenol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H27557.14
H27557.06
H27557.03
H27557.14EA
209
EUR
H27557.14
H27557.06
H27557.03
4-Brom-2-(5-isoxazolyl)phenol ≥97%
4-Brom-2-(5-isoxazolyl)phenol
Purity: 97%
CAS No.: 213690-27-8
Molecular Formula: C9H6BrNO2
Formula Weight: 240.05
Melting Point: 207-212°
MDL No.: MFCD00799493
Formel:
C₉H₆BrNO₂ Molecular Weight: 240,05 g/mol Schmelzpunkt: 207…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00799493 CAS-Nummer: 213690-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...