5-(3-Chlorphenyl)isoxazol

Lieferant: COMBI-BLOCKS

HC-6886-1G HC-6886-5G
COBBHC-6886-1GEA 98.3 EUR
COBBHC-6886-1G COBBHC-6886-5G
5-(3-Chlorphenyl)isoxazol
5-(3-Chlorophenyl)isoxazole
Formel: C₉H₆ClNO
MDL: MFCD03232871
CAS-Nummer: 7064-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR