3-(2-Chlorphenyl)-5-isoxazolcarbonsäure

Lieferant: BIONET RESEARCH

11L-369S.500 11L-369S.1 11L-369S.5 11L-369S.10
BIOT11L-369S.500EA 0 EUR
BIOT11L-369S.500 BIOT11L-369S.1 BIOT11L-369S.5 BIOT11L-369S.10
3-(2-Chlorphenyl)-5-isoxazolcarbonsäure
3-(2-Chlorophenyl)-5-isoxazolecarboxylic acid
Formel: C₁₀H₆ClNO₃
MDL: MFCD01935954
CAS-Nummer: 338982-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR