3-(4-Methoxyphenyl)-5-isoxazolcarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-(4-Methoxyphenyl)isoxazole-5-carboxylic acid

11L-327S.1 11L-327S.5 11L-327S.500
BIOT11L-327S.1EA 0 EUR
BIOT11L-327S.1 BIOT11L-327S.5 BIOT11L-327S.500
3-(4-Methoxyphenyl)-5-isoxazolcarbonsäure
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid
Formel: C₁₁H₉NO₄
Molecular Weight: 219,1900 g/mol
MDL: MFCD04124288
CAS-Nummer: 618383-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR