Ethyl-2-brom-4-oxazolcarboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2-bromooxazole-4-carboxylate
HI-1196-5G
HI-1196-1G
COBBHI-1196-5GEA
462
EUR
COBBHI-1196-5G
COBBHI-1196-1G
Ethyl-2-brom-4-oxazolcarboxylat
Ethyl-2-brom-4-oxazolcarboxylat
Formel:
C₆H₆BrNO₃ Molecular Weight: 220,0207 g/mol |
MDL:
MFCD09801023 CAS-Nummer: 460081-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...