Ethyl-2-(tert-butoxycarbonylamino)oxazol-5-carboxylat

Lieferant: COMBI-BLOCKS

HI-1323-5G HI-1323-1G
COBBHI-1323-5GEA 0 EUR
COBBHI-1323-5G COBBHI-1323-1G
Ethyl-2-(tert-butoxycarbonylamino)oxazol-5-carboxylat
Ethyl-2-(tert-butoxycarbonylamino)oxazol-5-carboxylat
Formel: C₁₁H₁₆N₂O₅
MDL: MFCD09475909
CAS-Nummer: 941294-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR