Ethyl-4-oxazolcarboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl oxazole-4-carboxylate
HI-1273-5G
HI-1273-1G
COBBHI-1273-5GEA
50.9
EUR
COBBHI-1273-5G
COBBHI-1273-1G
Ethyl-4-oxazolcarboxylat
Ethyl-4-oxazolcarboxylat
Formel:
C₆H₇NO₃ Molecular Weight: 141,1300 g/mol |
MDL:
MFCD04114940 CAS-Nummer: 23012-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...