Ethyl-4-oxazolcarboxylat

Lieferant: CHEMPUR

Synonyme: Ethyl oxazole-4-carboxylate

56-0023.1 56-0023.5
CHMP56-0023.1EA 0 EUR
CHMP56-0023.1 CHMP56-0023.5
Ethyl-4-oxazolcarboxylat
Ethyl-4-oxazolcarboxylat
Formel: C₆H₇NO₃
Molecular Weight: 141,1300 g/mol
MDL: MFCD04114940
CAS-Nummer: 23012-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR