Ethyl-5-oxazolcarboxylat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl oxazole-5-carboxylate
H32924.14
H32924.06
H32924.14EA
224
EUR
H32924.14
H32924.06
Ethyl-5-oxazolcarboxylat ≥98%
Ethyl-5-oxazolcarboxylat
Purity: 98%
CAS No.: 118994-89-1
Molecular Formula: C6H7NO3
Formula Weight: 141.13
Refractive Index: 1.4690
Boiling Point: 60°/2.25mm
MDL No.: MFCD04114930
Formel:
C₆H₇NO₃ Molecular Weight: 141.13 g/mol Siedepunkt: 60 °C (2.25 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114930 CAS-Nummer: 118994-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...