Ethyl-5-oxazolcarboxylat
Lieferant: CHEMPUR
Synonyme:
Ethyl oxazole-5-carboxylate
56-0007.1
56-0007.5
CHMP56-0007.1EA
0
EUR
CHMP56-0007.1
CHMP56-0007.5
Ethyl-5-oxazolcarboxylat
Ethyl-5-oxazolcarboxylat
Formel:
C₆H₇NO₃ Molecular Weight: 141.13 g/mol |
MDL:
MFCD04114930 CAS-Nummer: 118994-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...