Ethyl-2-amino-4-oxazolcarboxylat 95%

Lieferant: Thermo Scientific

Synonyme: Ethyl 2-aminooxazole-4-carboxylate

H32542.03 H32542.06
H32542.03EA 48.3 EUR
H32542.03 H32542.06
Ethyl-2-amino-4-oxazolcarboxylat 95%
Ethyl-2-amino-4-oxazolcarboxylat

Purity: 95% CAS No.: 177760-52-0 Molecular Formula: C6H8N2O3 Formula Weight: 156.14 Melting Point: 124-128° MDL No.: MFCD06659907

Formel: C₆H₈N₂O₃
Molecular Weight: 156,14 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659907
CAS-Nummer: 177760-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR