4-Oxazolcarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: Oxazole-4-carboxylic acid

HI-1274-5G HI-1274-1G
COBBHI-1274-5GEA 153 EUR
COBBHI-1274-5G COBBHI-1274-1G
4-Oxazolcarbonsäure
4-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,0715 g/mol
MDL: MFCD06797151
CAS-Nummer: 23012-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR