5-Oxazolcarbonsäure

Lieferant: CHEMPUR

Synonyme: Oxazole-5-carboxylic acid

56-0008.1 56-0008.5
CHMP56-0008.1EA 0 EUR
CHMP56-0008.1 CHMP56-0008.5
5-Oxazolcarbonsäure
5-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD04114931
CAS-Nummer: 118994-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR