2-Amino-4-oxazolcarbonsäure

Lieferant: COMBI-BLOCKS

HI-1515-5G HI-1515-1G
COBBHI-1515-5GEA 1140 EUR
COBBHI-1515-5G COBBHI-1515-1G
2-Amino-4-oxazolcarbonsäure
2-Amino-4-oxazolecarboxylic acid
Formel: C₄H₄N₂O₃
MDL: MFCD09864918
CAS-Nummer: 944900-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR