2-Amino-5-oxazolcarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Aminooxazole-5-carboxylic acid

HI-1433-5G HI-1433-1G
COBBHI-1433-5GEA 836 EUR
COBBHI-1433-5G COBBHI-1433-1G
2-Amino-5-oxazolcarbonsäure
2-Amino-5-oxazolecarboxylic acid
Formel: C₄H₄N₂O₃
Molecular Weight: 128,862 g/mol
MDL: MFCD11504937
CAS-Nummer: 881637-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR