2-Aminooxazol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Oxazol-2-amine
H34360.03
H34360.06
H34360.03EA
132
EUR
H34360.03
H34360.06
2-Aminooxazol 97%
2-Aminooxazole
Formel:
C₃H₄N₂O Molecular Weight: 84.10 g/mol Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07364485 CAS-Nummer: 4570-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...