5-(Ethylthio)-1H-tetrazol (ETT)

Lieferant: FLUOROCHEM

Synonyme: 5-Ethyltio-1H-Tetrazole

002927-1G F002927-5G
FLUO002927-1GEA 34.1 EUR
FLUO002927-1G FLUOF002927-5G
5-(Ethylthio)-1H-tetrazol (ETT)
5-(Ethylthio)-1H-tetrazol (ETT)
Formel: C₃H₆N₄S
Molecular Weight: 130,17 g/mol
MDL: MFCD00068733
CAS-Nummer: 89797-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR