5-Mercapto-1-methyl-1H-tetrazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Mercapto-1-methyltetrazole

B20443.06 B20443.14
B20443.06EA 38.2 EUR
B20443.06 B20443.14
5-Mercapto-1-methyl-1H-tetrazol ≥98%
5-Mercapto-1-methyl-1H-tetrazole

Purity: 98% CAS No.: 13183-79-4 Molecular Formula: C2H4N4S Formula Weight: 116.14 Melting Point: 125-128° MDL No.: MFCD00037317

Formel: C₂H₄N₄S
Molecular Weight: 116,14 g/mol
Schmelzpunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037317
CAS-Nummer: 13183-79-4
EINECS: 236-132-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR