5-(4-Methoxyphenyl)-1H-tetrazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-(4-Methoxyphenyl)-2H-tetrazole

L17210.03 L17210.06
L17210.03EA 71.9 EUR
L17210.03 L17210.06
5-(4-Methoxyphenyl)-1H-tetrazol ≥98%
5-(4-Methoxyphenyl)-1H-tetrazole

Purity: 98% CAS No.: 6926-51-8 Molecular Formula: C8H8N4O Formula Weight: 176.18 Melting Point: 235-238° Beilstein Reference: 152619

Formel: C₈H₈N₄O
Molecular Weight: 176,18 g/mol
Schmelzpunkt: 235…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00991875
CAS-Nummer: 6926-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR