Ethylthiazol-4-carboxylat

Lieferant: CHEMPUR

90-0009.1 90-0009.5
CHMP90-0009.1EA 0 EUR
CHMP90-0009.1 CHMP90-0009.5
Ethylthiazol-4-carboxylat
Ethylthiazol-4-carboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.1900 g/mol
MDL: MFCD03788566
CAS-Nummer: 14527-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR