Methylthiazol-4-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-thiazolecarboxylate

HI-1150-5G HI-1150-25G HI-1150-1G
COBBHI-1150-5GEA 88.6 EUR
COBBHI-1150-5G COBBHI-1150-25G COBBHI-1150-1G
Methylthiazol-4-carboxylat
Methylthiazol-4-carboxylat
Formel: C₅H₅NO₂S
Molecular Weight: 143.1600 g/mol
MDL: MFCD06410940
CAS-Nummer: 59418-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR