Ethylthiazol-5-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 5-thiazolecarboxylate

HI-1149-1G HI-1149-5G
COBBHI-1149-1GEA 49.7 EUR
COBBHI-1149-1G COBBHI-1149-5G
Ethylthiazol-5-carboxylat
Ethylthiazol-5-carboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.1900 g/mol
MDL: MFCD06205069
CAS-Nummer: 32955-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR