2-Amino-5-thiazolcarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Aminothiazole-5-carboxylic acid

HI-1130-5G HI-1130-1G
COBBHI-1130-5GEA 35.8 EUR
COBBHI-1130-5G COBBHI-1130-1G
2-Amino-5-thiazolcarbonsäure
2-Amino-5-thiazolecarboxylic acid
Formel: C₄H₄N₂O₂S
Molecular Weight: 144,1500 g/mol
MDL: MFCD06203554
CAS-Nummer: 40283-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR