Ethyl-2-methylthiazol-4-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2-methylthiazole-4-carboxylate
OS-7705-5G
OS-7705-25G
OS-7705-1G
COBBOS-7705-5GEA
88.6
EUR
COBBOS-7705-5G
COBBOS-7705-25G
COBBOS-7705-1G
Ethyl-2-methylthiazol-4-carboxylat
Ethyl-2-methylthiazol-4-carboxylat
Formel:
C₇H₉NO₂S Molecular Weight: 171.22 g/mol |
MDL:
MFCD00156148 CAS-Nummer: 6436-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...