5-(3-Chlorphenyl)oxazol

Lieferant: Thermo Scientific

SEW00955.1 SEW00955.10
MAYBSEW00955.1EA 0 EUR
MAYBSEW00955.1 MAYBSEW00955.10
5-(3-Chlorphenyl)oxazol
5-(3-Chlorophenyl)oxazole
Formel: C₉H₆ClNO
Molecular Weight: 179,6000 g/mol
MDL: MFCD00084974
CAS-Nummer: 89808-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR