Ethyl-5-(4-chlorphenyl)-4-oxazolcarboxylat

Lieferant: COMBI-BLOCKS

HC-6231-5G HC-6231-1G
COBBHC-6231-5GEA 2070 EUR
COBBHC-6231-5G COBBHC-6231-1G
Ethyl-5-(4-chlorphenyl)-4-oxazolcarboxylat
Ethyl-5-(4-chlorphenyl)-4-oxazolcarboxylat
Formel: C₁₂H₁₀ClNO₃
MDL: MFCD00085064
CAS-Nummer: 127919-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR