5-(2,3-Dichlorphenyl)-4-oxazolcarbonsäure
Lieferant: COMBI-BLOCKS
HC-6237-5G
HC-6237-1G
COBBHC-6237-5GEA
915
EUR
COBBHC-6237-5G
COBBHC-6237-1G
5-(2,3-Dichlorphenyl)-4-oxazolcarbonsäure
5-(2,3-Dichlorophenyl)-4-oxazolecarboxylic acid
Formel:
C₁₀H₅Cl₂NO₃ |
MDL:
MFCD07376179 CAS-Nummer: 951885-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...