5-(2,4-Dichlorphenyl)-4-oxazolcarbonsäure

Lieferant: COMBI-BLOCKS

HC-6238-5G HC-6238-1G
COBBHC-6238-5GEA 595 EUR
COBBHC-6238-5G COBBHC-6238-1G
5-(2,4-Dichlorphenyl)-4-oxazolcarbonsäure
5-(2,4-Dichlorophenyl)-4-oxazolecarboxylic acid
Formel: C₁₀H₅Cl₂NO₃
MDL: MFCD01934490
CAS-Nummer: 255876-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR