5-(4-Chlorphenyl)-4-oxazolcarbonsäure

Lieferant: CHEMPUR

FL21359.POR 021359-100MG
CHMPFL21359.POREA 0 EUR
CHMPFL21359.POR FLUO021359-100MG
5-(4-Chlorphenyl)-4-oxazolcarbonsäure
5-(4-Chlorophenyl)-4-oxazolecarboxylic acid
Formel: C₁₀H₆ClNO₃
MDL: MFCD00105450
CAS-Nummer: 143659-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR