Ethyl-5-(3-nitrophenyl)-4-oxazolcarboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 5-(3-nitrophenyl)oxazole-4-carboxylate
HC-6227-5G
HC-6227-1G
COBBHC-6227-5GEA
451
EUR
COBBHC-6227-5G
COBBHC-6227-1G
Ethyl-5-(3-nitrophenyl)-4-oxazolcarboxylat
Ethyl-5-(3-nitrophenyl)-4-oxazolcarboxylat
Formel:
C₁₂H₁₀N₂O₅ Molecular Weight: 262.218 g/mol |
MDL:
MFCD09800990 CAS-Nummer: 916674-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...