Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 5-(4-nitrophenyl)oxazole-4-carboxylate
HC-6230-5G
HC-6230-1G
COBBHC-6230-5GEA
374
EUR
COBBHC-6230-5G
COBBHC-6230-1G
Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Formel:
C₁₂H₁₀N₂O₅ Molecular Weight: 262.2200 g/mol |
MDL:
MFCD02179429 CAS-Nummer: 72030-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...