Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat

Lieferant: CHEMPUR

Synonyme: Ethyl 5-(4-nitrophenyl)oxazole-4-carboxylate

56-0034.1 56-0034.5
CHMP56-0034.1EA 0 EUR
CHMP56-0034.1 CHMP56-0034.5
Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Ethyl-5-(4-nitrophenyl)-4-oxazolcarboxylat
Formel: C₁₂H₁₀N₂O₅
Molecular Weight: 262.2200 g/mol
MDL: MFCD02179429
CAS-Nummer: 72030-87-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR