5-(3-Nitrophenyl)-4-oxazolcarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-(3-Nitrophenyl)-1,3-oxazole-4-carboxylic acid
HC-6236-5G
HC-6236-1G
COBBHC-6236-5GEA
595
EUR
COBBHC-6236-5G
COBBHC-6236-1G
5-(3-Nitrophenyl)-4-oxazolcarbonsäure
5-(3-Nitrophenyl)-4-oxazolecarboxylic acid
Formel:
C₁₀H₆N₂O₅ |
MDL:
MFCD09800992 CAS-Nummer: 951885-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...