2-(4-Fluorphenyl)oxazol

Lieferant: COMBI-BLOCKS

HC-3306-1G HC-3306-5G
COBBHC-3306-1GEA 0 EUR
COBBHC-3306-1G COBBHC-3306-5G
2-(4-Fluorphenyl)oxazol
2-(4-Fluorophenyl)oxazole
Formel: C₉H₆FNO
MDL: MFCD08234435
CAS-Nummer: 885268-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR