5-(4-Fluorphenyl)oxazol

Lieferant: Thermo Scientific

SEW00971.1 SEW00971.10
MAYBSEW00971.1EA 0 EUR
MAYBSEW00971.1 MAYBSEW00971.10
5-(4-Fluorphenyl)oxazol
5-(4-Fluorophenyl)oxazole
Formel: C₉H₆FNO
MDL: MFCD00085087
CAS-Nummer: 128101-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR