5-(2-Fluorphenyl)-4-oxazolcarbonsäure

Lieferant: COMBI-BLOCKS

HC-6232-1G HC-6232-5G
COBBHC-6232-1GEA 557 EUR
COBBHC-6232-1G COBBHC-6232-5G
5-(2-Fluorphenyl)-4-oxazolcarbonsäure
5-(2-Fluorophenyl)-4-oxazolecarboxylic acid
Formel: C₁₀H₆FNO₃
MDL: MFCD07376192
CAS-Nummer: 347187-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR