N,N-Dimethyl-1-dodecylamin-N-oxid 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-ethyl-N-oxido-dodecan-1-amine, LDAO, Lauryldimethylamine oxide, N,N-Dimethyldodecan-1-amine oxide, Lauramine oxide, N,N-Dimethyl-N-dodecylamine N-oxide

H26065.14 H26065.06
H26065.14EA 534 EUR
H26065.14 H26065.06
N,N-Dimethyl-1-dodecylamin-N-oxid 95%
N,N-Dimethyl-1-dodecylamin-N-oxid

Purity: 95% CAS No.: 1643-20-5 Molecular Formula: C14H31NO Formula Weight: 229.40 Melting Point: 120-123° Beilstein Reference: 1769927 UN No.: 3259 MDL No.: MFCD00002049

Formel: C₁₄H₃₁NO
Molecular Weight: 229,40 g/mol
Schmelzpunkt: 120…123 °C
Dichte: 0,996
Lagertemperatur: Kühlschrank
MDL: MFCD00002049
CAS-Nummer: 1643-20-5
EINECS: 216-700-6
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR